Assay Details
Target Gene Details
Entrez Gene ID: | 26 |
Gene Name: | amine oxidase, copper containing 1 |
Gene Aliases: |
ABP, ABP1, DAO, DAO1, KAO |
Location: |
Chr.7:150824875-150861291 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AOC1 | XM_017011944.1 | XP_016867433.1 | ||
XM_017011945.1 | XP_016867434.1 | |||
BX648159.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1030109 | Chr.7:150615828 - 151003922 on Build GRCh38 | Gain | LOC105375566 GIMAP1 GIMAP5 GIMAP6 GIMAP1-GIMAP5 TMEM176B GIMAP2 NOS3 KCNH2 AOC1 TMEM176A |
esv2735390 | Chr.7:150829998 - 150842197 on Build GRCh38 | Deletion | AOC1 |
nsv608990 | Chr.7:150264185 - 151010400 on Build GRCh38 | Gain | REPIN1 LOC105375566 GIMAP8 GIMAP5 LOC107986860 LOC728743 TMEM176B GIMAP2 ZNF775 GIMAP7 RARRES2 KCNH2 AOC1 ACTR3C GIMAP4 GIMAP1 GIMAP6 GIMAP1-GIMAP5 NOS3 LRRC61 ZBED6CL LINC00996 TMEM176A |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |