Assay Details
Target Gene Details
Entrez Gene ID: | 644 |
Gene Name: | biliverdin reductase A |
Gene Aliases: |
BLVR, BVR, BVRA |
Location: |
Chr.7:43758153-43807342 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BLVRA | NM_000712.3 | NP_000703.2 | ||
NM_001253823.1 | NP_001240752.1 | |||
XM_011515474.2 | XP_011513776.1 | |||
XM_017012520.1 | XP_016868009.1 | |||
AK291862.1 | ||||
BC005902.1 | AAH05902.1 | |||
BC008456.1 | AAH08456.1 | |||
BM710258.1 | ||||
U34877.1 | AAC35588.1 | |||
X93086.1 | CAA63635.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv606745 | Chr.7:43652117 - 43787815 on Build GRCh38 | Loss | BLVRA COA1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |