Assay Details
Target Gene Details
Entrez Gene ID: | 2050 |
Gene Name: | EPH receptor B4 |
Gene Aliases: |
HTK, MYK1, TYRO11 |
Location: |
Chr.7:100802565-100827521 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EPHB4 | NM_004444.4 | NP_004435.3 | ||
XM_017011816.1 | XP_016867305.1 | |||
AB209644.1 | BAD92881.1 | |||
AK301224.1 | ||||
AY056048.1 | AAL14195.1 | |||
BC004264.1 | AAH04264.1 | |||
BC052804.1 | AAH52804.1 | |||
U07695.1 | AAA20598.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2759547 | Chr.7:100757867 - 101105965 on Build GRCh38 | Gain+Loss | UFSP1 TRIP6 SLC12A9 EPHB4 TRIM56 MUC3A MUC17 LOC102724094 MUC12 ACHE ZAN MIR6875 SRRT |
nsv470381 | Chr.7:100642568 - 100834358 on Build GRCh38 | Loss | EPO ACTL6B TFR2 GNB2 POP7 LOC105375429 EPHB4 ZAN GIGYF1 |
esv2734888 | Chr.7:99975623 - 101290285 on Build GRCh38 | Deletion | UFSP1 MIR4653 VGF LRCH4 TRIM56 MUC3A COPS6 C7orf43 AP4M1 MOSPD3 NYAP1 PVRIG2P LAMTOR4 MBLAC1 SERPINE1 ZAN TSC22D4 MIR6875 MIR25 ZKSCAN1 PPP1R35 POP7 ZNF3 SAP25 SLC12A9 NAT16 TAF6 AZGP1P1 PLOD3 ZSCAN21 LOC105375429 MIR93 PCOLCE-AS1 SRRT MIR106B GATS GNB2 ZASP TRIP6 GIGYF1 MCM7 PILRA PVRIG ACTL6B STAG3L5P-PVRIG2P-PILRB MOGAT3 CLDN15 AGFG2 FBXO24 MEPCE SPDYE3 AP1S1 PCOLCE AZGP1 GPC2 STAG3L5P EPHB4 PILRB MUC17 LOC102724094 MIR6840 C7orf61 STAG3 ZNHIT1 MUC12 EPO CNPY4 ACHE PMS2P1 TFR2 ZCWPW1 FIS1 MIR4658 GAL3ST4 |
nsv607965 | Chr.7:100776465 - 100899511 on Build GRCh38 | Loss | ACHE UFSP1 TRIP6 SLC12A9 EPHB4 ZAN MIR6875 SRRT |
nsv1020837 | Chr.7:100715499 - 100911015 on Build GRCh38 | Loss | EPO ACHE UFSP1 TRIP6 SLC12A9 EPHB4 ZAN MIR6875 SRRT |
nsv951388 | Chr.7:100741378 - 100838178 on Build GRCh38 | Deletion | EPHB4 ZAN |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |