Assay Details
Target Gene Details
Entrez Gene ID: | 285965 |
Gene Name: | EPHA1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.7:143407813-143523449 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EPHA1-AS1 | NR_033897.1 | |||
AL833583.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv608682 | Chr.7:143444382 - 144177097 on Build GRCh38 | Gain | CTAGE15 LOC101928422 TCAF2P1 TAS2R41 OR2F1 TCAF1 LOC154761 CTAGE6 OR2A12 TAS2R60 OR2A14 EPHA1-AS1 OR2A5 TCAF2 OR6B1 OR2F2 OR2A2 OR2A25 |