Assay Details
Target Gene Details
Entrez Gene ID: | 84068 |
Gene Name: | solute carrier family 10 member 7 |
Gene Aliases: |
C4orf13, P7 |
Location: |
Chr.4:146253981-146522351 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC10A7 | NM_001029998.5 | NP_001025169.1 | ||
NM_001300842.2 | NP_001287771.1 | |||
NM_001317816.1 | NP_001304745.1 | |||
NR_133924.1 | ||||
XM_011532311.2 | XP_011530613.1 | |||
XM_011532313.2 | XP_011530615.1 | |||
XM_011532314.2 | XP_011530616.1 | |||
AK075364.1 | BAC11571.1 | |||
AK126000.1 | ||||
AK172727.1 | ||||
AK291046.1 | ||||
AY346324.1 | AAQ84722.1 | |||
BC150308.1 | ||||
CR933647.1 | CAI45948.1 | |||
CR936751.1 | ||||
DQ122860.1 | AAZ32256.1 | |||
DQ122861.1 | AAZ32257.1 | |||
DQ871036.1 | ABI31650.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv461675 | Chr.4:146195712 - 146847054 on Build GRCh38 | Gain | LOC345051 POU4F2 TTC29 LSM6 MIR7849 SLC10A7 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |