Assay Details
Target Gene Details
Entrez Gene ID: | 836 |
Gene Name: | caspase 3 |
Gene Aliases: |
CPP32, CPP32B, SCA-1 |
Location: |
Chr.4:184627696-184649475 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CASP3 | NM_004346.3 | NP_004337.2 | ||
NM_032991.2 | NP_116786.1 | |||
XM_011532301.1 | XP_011530603.1 | |||
AK291337.1 | ||||
AU127028.1 | ||||
AU133051.1 | ||||
BC016926.2 | AAH16926.1 | |||
BX647609.1 | CAI46084.1 | |||
U13737.1 | AAA65015.1 | |||
U13738.1 | AAB60355.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2763423 | Chr.4:184642465 - 184688651 on Build GRCh38 | Loss | CASP3 PRIMPOL |
nsv830168 | Chr.4:184626297 - 184801578 on Build GRCh38 | Loss | CENPU SLED1 CASP3 ACSL1 PRIMPOL |
nsv1026074 | Chr.4:184384963 - 184780663 on Build GRCh38 | Gain | CENPU IRF2 LVCAT8 CASP3 ACSL1 PRIMPOL |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |