Assay Details
Target Gene Details
Entrez Gene ID: | 407738 |
Gene Name: | family with sequence similarity 19 member A1, C-C motif chemokine like |
Gene Aliases: |
TAFA-1, TAFA1 |
Location: |
Chr.3:67991592-68546056 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FAM19A1 | NM_001252216.1 | NP_001239145.1 | ||
NM_213609.3 | NP_998774.2 | |||
XM_017006386.1 | XP_016861875.1 | |||
XM_017006387.1 | XP_016861876.1 | |||
XM_017006388.1 | XP_016861877.1 | |||
XM_017006389.1 | XP_016861878.1 | |||
XM_017006390.1 | XP_016861879.1 | |||
AK289668.1 | ||||
AK290446.1 | ||||
AL713702.1 | CAD28501.1 | |||
AY325114.1 | AAP92406.1 | |||
BC025746.1 | AAH25746.1 | |||
DA485098.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1003158 | Chr.3:66999779 - 68395539 on Build GRCh38 | Loss | FAM19A1 MIR4272 SUCLG2 SUCLG2-AS1 KBTBD8 |
esv999186 | Chr.3:68017838 - 68032369 on Build GRCh38 | Insertion | FAM19A1 |
nsv1013531 | Chr.3:67867347 - 68020883 on Build GRCh38 | Gain | FAM19A1 SUCLG2-AS1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |