Assay Details
Target Gene Details
Entrez Gene ID: | 54585 |
Gene Name: | leucine zipper transcription factor like 1 |
Gene Aliases: |
BBS17 |
Location: |
Chr.3:45823316-45916042 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LZTFL1 | NM_001276378.1 | NP_001263307.1 | ||
NM_001276379.1 | NP_001263308.1 | |||
XM_017006644.1 | XP_016862133.1 | |||
XM_017006645.1 | XP_016862134.1 | |||
AK093705.1 | ||||
AK303416.1 | ||||
DC410047.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834678 | Chr.3:45762802 - 45968764 on Build GRCh38 | Gain | CCR9 FYCO1 CXCR6 LZTFL1 SLC6A20 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |