Assay Details
Target Gene Details
Entrez Gene ID: | 10063 |
Gene Name: | COX17, cytochrome c oxidase copper chaperone |
Gene Aliases: |
- |
Location: |
Chr.3:119659975-119677441 on Build GRCh38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
COX17 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1014394 | Chr.3:119641659 - 119693428 on Build GRCh38 | Loss | COX17 POPDC2 |
esv3568977 | Chr.3:119657752 - 119694492 on Build GRCh38 | Loss | COX17 POPDC2 |
nsv591390 | Chr.3:119665316 - 119740004 on Build GRCh38 | Gain | COX17 MAATS1 |