Assay Details
Target Gene Details
Entrez Gene ID: | 55825 |
Gene Name: | peroxisomal trans-2-enoyl-CoA reductase |
Gene Aliases: |
DCRRP, HPDHASE, HSA250303, PVIARL, SDR29C1, TERP |
Location: |
Chr.2:216029088-216081823 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PECR | NM_018441.5 | NP_060911.2 | ||
XM_011511478.2 | XP_011509780.1 | |||
AF212234.1 | AAK14920.1 | |||
AF232009.1 | AAF69798.1 | |||
AJ250303.1 | CAB89810.1 | |||
AK296208.1 | ||||
AK315795.1 | ||||
BC002529.2 | AAH02529.1 | |||
BU151224.1 | ||||
CR457145.1 | CAG33426.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1011506 | Chr.2:215969945 - 216310768 on Build GRCh38 | Gain | PKI55 PECR MARCH4 MREG XRCC5 TMEM169 |
nsv998837 | Chr.2:215982428 - 216143671 on Build GRCh38 | Gain | PECR MREG XRCC5 TMEM169 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |