Assay Details
Target Gene Details
Entrez Gene ID: | 101928371 |
Gene Name: | uncharacterized LOC101928371 |
Gene Aliases: |
- |
Location: |
Chr.2:88538720-88575610 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC101928371 | NR_110236.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv470476 | Chr.2:88384248 - 88829483 on Build GRCh38 | Gain | LOC101928403 RPIA LOC101928371 MIR4436A EIF2AK3 ANKRD36BP2 FOXI3 TEX37 |
dgv61n16 | Chr.2:87343737 - 90254660 on Build GRCh38 | Deletion | KRCC1 LOC107985911 EIF2AK3 LOC730268 IGK FABP1 MIR4780 RGPD2 SMYD1 MIR4436A FOXI3 TEX37 LOC101928403 MIR4435-1 RPIA IGKV2-29 IGKV2-24 IGKV1D-8 IGKV3-20 IGKV2-26 PLGLB2 IGKV3D-15 IGKV1D-13 LOC101927050 LOC101928371 THNSL2 LINC00152 IGKV1-5 ANKRD36BP2 IGKC |
dgv6947n54 | Chr.2:87815738 - 88850700 on Build GRCh38 | Gain | LOC101928403 KRCC1 RPIA EIF2AK3 FABP1 MIR4780 RGPD2 SMYD1 LOC101928371 THNSL2 MIR4436A ANKRD36BP2 FOXI3 TEX37 |
nsv1072005 | Chr.2:87360367 - 90256528 on Build GRCh38 | Deletion | KRCC1 LOC107985911 EIF2AK3 LOC730268 IGK FABP1 MIR4780 RGPD2 SMYD1 MIR4436A FOXI3 TEX37 LOC101928403 MIR4435-1 RPIA IGKV2-29 IGKV2-24 IGKV1D-8 IGKV3-20 IGKV2-26 PLGLB2 IGKV3D-15 IGKV1D-13 LOC101927050 LOC101928371 THNSL2 LINC00152 IGKV1-5 ANKRD36BP2 IGKC |
nsv1130752 | Chr.2:87360364 - 90256525 on Build GRCh38 | Deletion | KRCC1 LOC107985911 EIF2AK3 LOC730268 IGK FABP1 MIR4780 RGPD2 SMYD1 MIR4436A FOXI3 TEX37 LOC101928403 MIR4435-1 RPIA IGKV2-29 IGKV2-24 IGKV1D-8 IGKV3-20 IGKV2-26 PLGLB2 IGKV3D-15 IGKV1D-13 LOC101927050 LOC101928371 THNSL2 LINC00152 IGKV1-5 ANKRD36BP2 IGKC |
nsv582384 | Chr.2:87987149 - 88773214 on Build GRCh38 | Loss | LOC101928403 KRCC1 RPIA EIF2AK3 FABP1 MIR4780 RGPD2 SMYD1 LOC101928371 THNSL2 ANKRD36BP2 FOXI3 TEX37 |
nsv582386 | Chr.2:88035188 - 89208087 on Build GRCh38 | Loss | LOC101928403 KRCC1 RPIA IGKV2-24 EIF2AK3 IGKV3-20 IGKV2-26 IGK FABP1 MIR4780 SMYD1 LOC101928371 THNSL2 MIR4436A IGKV1-5 ANKRD36BP2 FOXI3 TEX37 IGKC |