Assay Details
Target Gene Details
Entrez Gene ID: | 2820 |
Gene Name: | glycerol-3-phosphate dehydrogenase 2 |
Gene Aliases: |
GDH2, GPDM, mGPDH |
Location: |
Chr.2:156435290-156586403 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GPD2 | NM_000408.4 | NP_000399.3 | ||
NM_001083112.2 | NP_001076581.2 | |||
XM_011510977.2 | XP_011509279.1 | |||
XM_017003830.1 | XP_016859319.1 | |||
XM_017003831.1 | XP_016859320.1 | |||
AB209399.1 | BAD92636.1 | |||
AK290057.1 | ||||
AK291065.1 | ||||
AK292817.1 | ||||
AK299645.1 | ||||
AL699359.1 | ||||
BC019874.1 | AAH19874.1 | |||
U12424.1 | AAA65701.1 | |||
U36310.1 | AAB60403.1 | |||
U79250.1 | AAB50200.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |