Assay Details
Target Gene Details
Entrez Gene ID: | 94101 |
Gene Name: | ORMDL sphingolipid biosynthesis regulator 1 |
Gene Aliases: |
- |
Location: |
Chr.2:189766149-189784371 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ORMDL1 | NM_001128150.1 | NP_001121622.1 | ||
NM_016467.4 | NP_057551.1 | |||
XM_005246970.2 | XP_005247027.1 | |||
XM_011512199.2 | XP_011510501.1 | |||
XM_011512200.2 | XP_011510502.1 | |||
XM_017005347.1 | XP_016860836.1 | |||
XM_017005348.1 | XP_016860837.1 | |||
AB064959.1 | BAC11710.1 | |||
AB064960.1 | BAC11711.1 | |||
AF151036.1 | AAF36122.1 | |||
AF395704.1 | AAM43503.1 | |||
AK074756.1 | BAC11184.1 | |||
AK075160.1 | BAC11441.1 | |||
AK313533.1 | ||||
BC005200.1 | AAH05200.1 | |||
CR936635.1 | CAI56776.1 | |||
DB122266.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |