Assay Details
Target Gene Details
Entrez Gene ID: | 57007 |
Gene Name: | atypical chemokine receptor 3 |
Gene Aliases: |
CMKOR1, CXC-R7, CXCR-7, CXCR7, GPR159, RDC-1, RDC1 |
Location: |
Chr.2:236537131-236582358 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ACKR3 | XM_005246098.3 | XP_005246155.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1005677 | Chr.2:236369512 - 236811991 on Build GRCh38 | Gain | ACKR3 LOC105373991 IQCA1 |
nsv997438 | Chr.2:236210757 - 237034862 on Build GRCh38 | Gain | ACKR3 LOC105373991 ASB18 IQCA1 |
nsv1000252 | Chr.2:235899999 - 238766417 on Build GRCh38 | Loss | RAMP1 COL6A3 MIR6811 RAB17 GBX2 LOC93463 ACKR3 LOC105373944 LOC101927958 LOC151174 AGAP1 ASB18 COPS8 LINC01107 ASB1 MLPH SCLY LOC643387 IQCA1 LOC105373958 UBE2F ESPNL UBE2F-SCLY LRRFIP1 ILKAP PER2 KLHL30 LOC105373991 ERFE TRAF3IP1 HES6 RBM44 PRLH |
nsv834576 | Chr.2:236466567 - 236654210 on Build GRCh38 | Gain | ACKR3 LOC105373991 IQCA1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |