Assay Details
Target Gene Details
Entrez Gene ID: | 9470 |
Gene Name: | eukaryotic translation initiation factor 4E family member 2 |
Gene Aliases: |
4E-LP, 4EHP, EIF4EL3, IF4e |
Location: |
Chr.2:232550587-232583645 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EIF4E2 | NM_001276336.1 | NP_001263265.1 | ||
NM_001276337.1 | NP_001263266.1 | |||
NM_001282958.1 | NP_001269887.1 | |||
NM_004846.3 | NP_004837.1 | |||
XM_005246975.3 | XP_005247032.1 | |||
XM_011512206.2 | XP_011510508.1 | |||
XM_017005377.1 | XP_016860866.1 | |||
AB209519.1 | BAD92756.1 | |||
AF038957.1 | AAC39871.1 | |||
AF047695.1 | AAC18565.1 | |||
AF068117.1 | AAC19374.1 | |||
AK022518.1 | ||||
AK303802.1 | ||||
BC005392.1 | AAH05392.1 | |||
BC005874.2 | AAH05874.1 | |||
BC021226.2 | AAH21226.1 | |||
BC021690.1 | AAH21690.1 | |||
BG913475.1 | ||||
CD684451.1 | ||||
DA992544.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv821918 | Chr.2:232319063 - 232560965 on Build GRCh38 | Gain | ECEL1 MIR5001 DIS3L2 ECEL1P2 EIF4E2 CHRND ALPI CHRNG ALPP TIGD1 ALPPL2 PRSS56 |
nsv3212 | Chr.2:232526220 - 232564112 on Build GRCh38 | Deletion | MIR5001 EIF4E2 CHRND CHRNG TIGD1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |