Assay Details
Target Gene Details
Entrez Gene ID: | 10458 |
Gene Name: | BAI1 associated protein 2 |
Gene Aliases: |
BAP2, FLAF3, IRSP53 |
Location: |
Chr.17:81035147-81117432 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv26223 | Chr.17:80659041 - 81736487 on Build GRCh38 | Gain+Loss | LOC107985021 C17orf89 LOC105371925 BAIAP2 SLC25A10 PDE6G AATK FSCN2 LOC105371921 FAAP100 RPTOR TEPSIN MIR3186 LOC400627 OXLD1 ACTG1 LOC100129503 MIR3065 LOC107985022 ARL16 MIR1250 TMEM105 MIR657 MIR4740 SLC38A10 TSPAN10 AATK-AS1 LINC00482 BAIAP2-AS1 BAHCC1 MIR6786 CEP131 LOC100130370 MRPL12 HGS CHMP6 NPLOC4 LOC101928855 MIR338 CCDC137 |
nsv833564 | Chr.17:81014713 - 81164054 on Build GRCh38 | Loss | MIR657 BAIAP2 BAIAP2-AS1 AATK MIR338 MIR3065 MIR1250 |
nsv470619 | Chr.17:81032233 - 81349843 on Build GRCh38 | Loss | C17orf89 MIR657 LOC105371925 SLC38A10 BAIAP2 AATK-AS1 LINC00482 BAIAP2-AS1 AATK CEP131 TEPSIN MIR338 MIR3065 MIR1250 TMEM105 |
nsv576169 | Chr.17:81030816 - 81043967 on Build GRCh38 | Loss | BAIAP2 BAIAP2-AS1 |
dgv3292n100 | Chr.17:81019582 - 81204920 on Build GRCh38 | Gain | MIR657 BAIAP2 AATK-AS1 BAIAP2-AS1 AATK MIR338 MIR3065 CEP131 MIR1250 |
nsv952388 | Chr.17:81034701 - 81137300 on Build GRCh38 | Deletion | MIR657 BAIAP2 BAIAP2-AS1 AATK MIR338 MIR3065 MIR1250 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |