Assay Details
Target Gene Details
Entrez Gene ID: | 10249 |
Gene Name: | glycine-N-acyltransferase |
Gene Aliases: |
ACGNAT, CAT, GAT |
Location: |
Chr.11:58708757-58731974 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GLYAT | NM_005838.3 | NP_005829.3 | ||
NM_201648.2 | NP_964011.2 | |||
XM_017017087.1 | XP_016872576.1 | |||
AB013093.1 | ||||
AB665285.1 | ||||
AF023466.1 | AAB81453.1 | |||
BC009785.1 | AAH09785.1 | |||
BG401676.1 | ||||
BG617408.1 | ||||
BP277146.1 | ||||
CB161978.1 | ||||
CR456927.1 | CAG33208.1 | |||
DR004166.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832168 | Chr.11:58626587 - 58797178 on Build GRCh38 | Gain | GLYAT LOC105369314 |
nsv1050013 | Chr.11:58627125 - 59453617 on Build GRCh38 | Gain | LOC101927204 OR5A2 LOC100287413 MPEG1 GLYAT OR5A1 GLYATL1 DTX4 GLYATL2 GLYATL1P1 FAM111B LOC105369314 GLYATL1P2 FAM111A OR5AN1 LOC283194 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |