Assay Details
Target Gene Details
Entrez Gene ID: | 377841 |
Gene Name: | ectonucleoside triphosphate diphosphohydrolase 8 |
Gene Aliases: |
E-NTPDase, GLSR2492, NTPDase-8, UNQ2492 |
Location: |
Chr.9:137434364-137441710 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ENTPD8 | NM_001033113.1 | NP_001028285.1 | ||
NM_198585.2 | NP_940987.2 | |||
XM_011518665.2 | XP_011516967.1 | |||
XM_011518666.2 | XP_011516968.1 | |||
XM_011518668.1 | XP_011516970.1 | |||
AY359088.1 | AAQ89446.1 | |||
BC141810.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv616021 | Chr.9:137434689 - 137452821 on Build GRCh38 | Loss |
![]() ![]() ![]() |
esv2657589 | Chr.9:137439793 - 137442640 on Build GRCh38 | Deletion |
![]() |
nsv951208 | Chr.9:137379349 - 137467948 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() |
nsv6771 | Chr.9:137412670 - 137444666 on Build GRCh38 | Insertion |
![]() ![]() ![]() |
esv2759721 | Chr.9:137303332 - 137447299 on Build GRCh38 | Loss |
![]() ![]() ![]() |
nsv524322 | Chr.9:136795692 - 137482216 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1075700 | Chr.9:137439847 - 137441448 on Build GRCh38 | Deletion |
![]() |
dgv4178n106 | Chr.9:137439748 - 137441448 on Build GRCh38 | Deletion |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)