Assay Details
Target Gene Details
Entrez Gene ID: | 5447 |
Gene Name: | cytochrome p450 oxidoreductase |
Gene Aliases: |
CPR, CYPOR, P450R |
Location: |
Chr.7:75915102-75986855 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
POR | NM_000941.2 | NP_000932.3 | ||
AK129978.1 | ||||
AK290529.1 | ||||
AK312543.1 | ||||
BP244929.1 | ||||
BX648619.1 | CAH56151.1 | |||
DA701694.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv528969 | Chr.7:75074053 - 76920728 on Build GRCh38 | Gain | MDH2 NSUN5P1 CCL24 PMS2P2 STAG3L1 SSC4D LOC107986711 TMEM120A NCF1C RHBDD2 GTF2IP1 PMS2P3 POMZP3 LOC541473 STYXL1 MIR4651 FDPSP2 DTX2 CCL26 POM121C PMS2P10 SNORA14A SPDYE5 LOC100133091 GTF2IP7 SRRM3 HIP1 LOC105375355 TRIM73 ZP3 YWHAG GTF2IRD2B POR LOC441259 HSPB1 SPDYE16 |
esv3613742 | Chr.7:75912784 - 76036530 on Build GRCh38 | Loss | TMEM120A SNORA14A STYXL1 POR MIR4651 |
esv3613743 | Chr.7:75917905 - 75929949 on Build GRCh38 | Loss | POR |
dgv120n16 | Chr.7:75440769 - 76641919 on Build GRCh38 | Deletion | MDH2 CCL24 SSC4D LOC107986711 TMEM120A RHBDD2 PMS2P3 POMZP3 STYXL1 MIR4651 FDPSP2 DTX2 CCL26 POM121C SNORA14A SPDYE5 LOC100133091 GTF2IP7 SRRM3 HIP1 LOC105375355 ZP3 YWHAG POR HSPB1 SPDYE16 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |