Assay Details
Target Gene Details
Entrez Gene ID: | 116449 |
Gene Name: | cytokine dependent hematopoietic cell linker |
Gene Aliases: |
MIST |
Location: |
Chr.4:10489899-10734853 on Build GRCh38 |
Assay Gene Location: | Within Intron 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CLNK | NM_052964.2 | NP_443196.2 | ||
XM_011513775.2 | XP_011512077.1 | |||
XM_017007684.1 | XP_016863173.1 | |||
AB032369.1 | BAA96241.1 | |||
AB110420.1 | BAC76765.1 | |||
AK093001.1 | ||||
BC137553.1 | ||||
BC137556.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv517971 | Chr.4:10397444 - 10891629 on Build GRCh38 | Gain | ZNF518B CLNK |
nsv593727 | Chr.4:10395703 - 10693367 on Build GRCh38 | Gain | ZNF518B CLNK |
esv3893877 | Chr.4:10411544 - 10500710 on Build GRCh38 | Loss | ZNF518B CLNK |
esv2759223 | Chr.4:10342075 - 10544176 on Build GRCh38 | Loss | ZNF518B CLNK |
esv3599624 | Chr.4:10476161 - 10518729 on Build GRCh38 | Loss | CLNK |
nsv1014335 | Chr.4:9435268 - 10687720 on Build GRCh38 | Gain | ZNF518B DRD5 MIR548I2 SLC2A9 MIR3138 OR7E85P WDR1 DEFB131 CLNK OR7E35P |
esv3893876 | Chr.4:10345427 - 10660766 on Build GRCh38 | Gain | ZNF518B CLNK |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |