Assay Details
Target Gene Details
Entrez Gene ID: | 375190 |
Gene Name: | family with sequence similarity 228 member B |
Gene Aliases: |
- |
Location: |
Chr.2:24076874-24169638 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FAM228B | NM_001291328.1 | NP_001278257.1 | ||
NR_111929.1 | ||||
BM722814.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv997445 | Chr.2:23357900 - 24374347 on Build GRCh38 | Gain | UBXN2A LOC105374325 KLHL29 MFSD2B TP53I3 ATAD2B PFN4 SF3B6 FAM228A FAM228B ITSN2 FKBP1B WDCP |
nsv997725 | Chr.2:24052436 - 25123406 on Build GRCh38 | Gain | PTRHD1 CENPO NCOA1 DNAJC27 ADCY3 TP53I3 DNAJC27-AS1 PFN4 SF3B6 FAM228A FAM228B ITSN2 EFR3B FKBP1B |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |