Assay Details
Target Gene Details
Entrez Gene ID: | 375056 |
Gene Name: | MIA family member 3, ER export factor |
Gene Aliases: |
ARNT, D320, TANGO, TANGO1, UNQ6077 |
Location: |
Chr.1:222617790-222668012 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MIA3 | NM_001300867.1 | NP_001287796.1 | ||
NM_001324062.1 | NP_001310991.1 | |||
NM_001324063.1 | NP_001310992.1 | |||
NM_001324064.1 | NP_001310993.1 | |||
NM_001324065.1 | NP_001310994.1 | |||
NM_198551.3 | NP_940953.2 | |||
XM_006711304.3 | XP_006711367.2 | |||
XM_011509513.2 | XP_011507815.2 | |||
XM_017001243.1 | XP_016856732.1 | |||
AK025122.1 | ||||
AK025506.1 | ||||
AK026489.1 | ||||
AK096526.1 | BAC04810.1 | |||
AK290335.1 | ||||
AK296712.1 | ||||
AX775845.1 | CAE11622.1 | |||
AY359091.1 | AAQ89449.1 | |||
D87742.1 | BAA13448.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |