Assay Details
Target Gene Details
Entrez Gene ID: | 10251 |
Gene Name: | sprouty RTK signaling antagonist 3 |
Gene Aliases: |
spry-3 |
Location: |
Chr.X:155612565-155782459 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SPRY3 | NM_001304990.1 | NP_001291919.1 | ||
DA102958.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv33784 | Chr.X:153962665 - 155699588 on Build GRCh38 | Gain+Loss | SNORA36A H2AFB1 LOC107987332 BRCC3 IKBKG G6PD CTAG2 FAM50A TMEM187 LOC105373386 OPN1MW3 CTAG1B MIR1184-3 DNASE1L1 UBL4A OPN1MW2 IRAK1 SNORA70 SPRY3 CLIC2 FAM223A PLXNA3 LAGE3 RPL10 TAZ MPP1 MIR6858 MTCP1 F8A2 MIR664B VBP1 ATP6AP1 HCFC1-AS1 H2AFB2 F8A1 LOC101927830 HCFC1 CH17-340M24.3 FLNA TMLHE-AS1 FAM3A MIR1184-2 H2AFB3 EMD MIR3202-2 FUNDC2 CTAG1A SNORA56 FAM223B MECP2 GAB3 OPN1LW SLC10A3 DKC1 CMC4 OPN1MW SMIM9 MIR1184-1 TEX28 F8 MIR718 GDI1 MIR3202-1 RAB39B F8A3 TMLHE TKTL1 |
nsv519485 | Chr.X:155507715 - 155699751 on Build GRCh38 | Loss | SPRY3 TMLHE |
nsv520699 | Chr.X:155545002 - 155699751 on Build GRCh38 | Gain | SPRY3 TMLHE |
nsv471607 | Chr.X:155652457 - 155812331 on Build GRCh38 | Gain+Loss | SPRY3 |
esv2657377 | Chr.X:155641695 - 155958434 on Build GRCh38 | Deletion | SPRY3 |
esv29972 | Chr.X:154781072 - 155699751 on Build GRCh38 | Gain | F8A1 H2AFB1 SPRY3 LOC101927830 BRCC3 CLIC2 CMC4 TMLHE-AS1 SMIM9 MIR1184-2 MIR1184-3 H2AFB3 MPP1 MIR1184-1 MTCP1 FUNDC2 F8A2 F8 VBP1 RAB39B F8A3 TMLHE H2AFB2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |