Assay Details
Target Gene Details
Entrez Gene ID: | 10251 |
Gene Name: | sprouty RTK signaling antagonist 3 |
Gene Aliases: |
spry-3 |
Location: |
Chr.X:155612565-155782459 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SPRY3 | NM_001304990.1 | NP_001291919.1 | ||
DA102958.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv33784 | Chr.X:153962665 - 155699588 on Build GRCh38 | Gain+Loss | TKTL1 RPL10 OPN1MW2 MPP1 MIR3202-1 LOC107987332 IRAK1 F8A2 MIR6858 VBP1 LOC101927830 G6PD HCFC1 OPN1MW SLC10A3 H2AFB2 IKBKG TAZ PLXNA3 OPN1MW3 SMIM9 EMD CTAG1A FLNA OPN1LW DKC1 CTAG2 DNASE1L1 CH17-340M24.3 CMC4 MTCP1 TEX28 MIR3202-2 FUNDC2 ATP6AP1 BRCC3 FAM3A SPRY3 RAB39B GDI1 SNORA36A MECP2 LOC105373386 SNORA70 MIR664B TMEM187 FAM223B MIR718 HCFC1-AS1 F8A3 CLIC2 CTAG1B FAM223A MIR1184-3 FAM50A GAB3 MIR1184-1 F8A1 SNORA56 TMLHE TMLHE-AS1 MIR1184-2 H2AFB3 LAGE3 UBL4A H2AFB1 F8 |
nsv519485 | Chr.X:155507715 - 155699751 on Build GRCh38 | Loss | TMLHE SPRY3 |
nsv520699 | Chr.X:155545002 - 155699751 on Build GRCh38 | Gain | TMLHE SPRY3 |
esv29972 | Chr.X:154781072 - 155699751 on Build GRCh38 | Gain | CMC4 MTCP1 MPP1 FUNDC2 MIR1184-3 BRCC3 SPRY3 RAB39B H2AFB2 MIR1184-1 F8A2 F8A1 SMIM9 TMLHE VBP1 TMLHE-AS1 LOC101927830 MIR1184-2 H2AFB3 H2AFB1 F8 F8A3 CLIC2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |