Assay Details
Target Gene Details
Entrez Gene ID: | 50814 |
Gene Name: | NAD(P) dependent steroid dehydrogenase-like |
Gene Aliases: |
H105E3, SDR31E1, XAP104 |
Location: |
Chr.X:152830967-152869363 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NSDHL | NM_001129765.1 | NP_001123237.1 | ||
NM_015922.2 | NP_057006.1 | |||
XM_011531178.2 | XP_011529480.1 | |||
XM_017029564.1 | XP_016885053.1 | |||
AK026549.1 | ||||
BC000245.1 | AAH00245.1 | |||
BC007816.2 | AAH07816.1 | |||
U47105.2 | AAC50558.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2758901 | Chr.X:152745001 - 152849208 on Build GRCh38 | Gain | CSAG3 NSDHL MAGEA6 MAGEA2 CETN2 |
nsv7158 | Chr.X:152841419 - 152868689 on Build GRCh38 | Insertion | NSDHL |
esv25868 | Chr.X:152816977 - 152847832 on Build GRCh38 | Gain | NSDHL CETN2 |
nsv470373 | Chr.X:152796870 - 152935948 on Build GRCh38 | Gain | NSDHL ZNF185 CETN2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |