Assay Details
Target Gene Details
Entrez Gene ID: | 10251 |
Gene Name: | sprouty RTK signaling antagonist 3 |
Gene Aliases: |
spry-3 |
Location: |
Chr.X:155612565-155782459 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SPRY3 | NM_001304990.1 | NP_001291919.1 | ||
DA102958.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv33784 | Chr.X:153962665 - 155699588 on Build GRCh38 | Gain+Loss | SNORA70 F8 MIR3202-1 FUNDC2 H2AFB3 LOC107987332 CTAG1B H2AFB1 OPN1MW RAB39B FAM223A GDI1 TEX28 SLC10A3 LOC105373386 TMLHE-AS1 FAM50A MTCP1 SNORA56 H2AFB2 ATP6AP1 CH17-340M24.3 EMD OPN1LW TMEM187 MPP1 G6PD LOC101927830 CMC4 TKTL1 RPL10 MIR6858 BRCC3 OPN1MW2 CTAG2 PLXNA3 MIR1184-1 MIR1184-3 VBP1 HCFC1-AS1 SNORA36A F8A2 CLIC2 F8A3 TMLHE FAM3A FAM223B F8A1 HCFC1 FLNA UBL4A IRAK1 DNASE1L1 TAZ MIR3202-2 MIR1184-2 MECP2 LAGE3 MIR718 GAB3 MIR664B OPN1MW3 SMIM9 CTAG1A SPRY3 IKBKG DKC1 |
nsv519485 | Chr.X:155507715 - 155699751 on Build GRCh38 | Loss | TMLHE SPRY3 |
nsv520699 | Chr.X:155545002 - 155699751 on Build GRCh38 | Gain | TMLHE SPRY3 |
esv29972 | Chr.X:154781072 - 155699751 on Build GRCh38 | Gain | TMLHE-AS1 MTCP1 F8 H2AFB2 FUNDC2 F8A1 MIR1184-1 MIR1184-3 VBP1 MPP1 H2AFB3 LOC101927830 H2AFB1 RAB39B F8A2 MIR1184-2 CLIC2 CMC4 F8A3 TMLHE SMIM9 BRCC3 SPRY3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |