Assay Details
Target Gene Details
Entrez Gene ID: | 55217 |
Gene Name: | trimethyllysine hydroxylase, epsilon |
Gene Aliases: |
AUTSX6, BBOX2, TMLD, TMLH, TMLHED, XAP130 |
Location: |
Chr.X:155489007-155612961 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TMLHE | NM_001184797.1 | NP_001171726.1 | ||
NM_018196.3 | NP_060666.1 | |||
XM_017029620.1 | XP_016885109.1 | |||
AF373407.1 | AAL01871.1 | |||
AK001589.1 | BAA91775.1 | |||
AK291694.1 | ||||
BC025269.1 | AAH25269.1 | |||
DA323339.1 | ||||
DB174485.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv510869 | Chr.X:155552481 - 155600316 on Build GRCh38 | Deletion |
![]() |
esv33784 | Chr.X:153962665 - 155699588 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv519485 | Chr.X:155507715 - 155699751 on Build GRCh38 | Loss |
![]() ![]() |
nsv7162 | Chr.X:155545842 - 155591711 on Build GRCh38 | Deletion |
![]() |
nsv520699 | Chr.X:155545002 - 155699751 on Build GRCh38 | Gain |
![]() ![]() |
esv3575078 | Chr.X:155556315 - 155574027 on Build GRCh38 | Loss |
![]() |
nsv511652 | Chr.X:155551462 - 155586778 on Build GRCh38 | Loss |
![]() |
esv29972 | Chr.X:154781072 - 155699751 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv24821 | Chr.X:155552998 - 155574053 on Build GRCh38 | Gain+Loss |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)