Assay Details
Target Gene Details
Entrez Gene ID: | 10587 |
Gene Name: | thioredoxin reductase 2 |
Gene Aliases: |
SELZ, TR, TR-BETA, TR3, TRXR2 |
Location: |
Chr.22:19875518-19941992 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TXNRD2 | NM_006440.4 | NP_006431.2 | ||
AB019694.1 | BAA77601.2 | |||
AB019695.1 | BAA77602.2 | |||
AB209724.1 | BAD92961.1 | |||
AF044212.1 | AAD25167.1 | |||
AF106697.1 | AAD19597.1 | |||
AF166126.1 | AAF21431.1 | |||
AF166127.1 | AAF21432.1 | |||
AF171054.1 | AAD51324.1 | |||
AF201385.1 | AAG47635.1 | |||
AK094698.1 | ||||
AK097708.1 | ||||
BC007489.2 | AAH07489.3 | |||
BC117354.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834129 | Chr.22:19705617 - 19919068 on Build GRCh38 | Loss | GP1BB C22orf29 TBX1 TXNRD2 GNB1L SEPT5-GP1BB SEPT5 |
esv3893434 | Chr.22:19470889 - 20343372 on Build GRCh38 | Gain | UFD1L TRMT2A TANGO2 TBX1 ZDHHC8 MIR1286 MIR4761 LINC00896 CCDC188 DGCR8 LOC101927859 TXNRD2 LOC440792 SEPT5-GP1BB LINC00895 LOC284865 CLDN5 GP1BB C22orf29 CDC45 MIR3618 ARVCF MIR1306 GNB1L SEPT5 MIR185 DGCR6L RTN4R RANBP1 MIR6816 COMT |
nsv828939 | Chr.22:19696032 - 20324465 on Build GRCh38 | Loss | TRMT2A TANGO2 TBX1 ZDHHC8 MIR1286 MIR4761 LINC00896 CCDC188 DGCR8 TXNRD2 LOC440792 SEPT5-GP1BB LOC284865 GP1BB C22orf29 MIR3618 ARVCF MIR1306 GNB1L SEPT5 MIR185 DGCR6L RTN4R RANBP1 MIR6816 COMT |
nsv828938 | Chr.22:19691069 - 20324465 on Build GRCh38 | Gain | TRMT2A TANGO2 TBX1 ZDHHC8 MIR1286 MIR4761 LINC00896 CCDC188 DGCR8 TXNRD2 LOC440792 SEPT5-GP1BB LOC284865 GP1BB C22orf29 MIR3618 ARVCF MIR1306 GNB1L SEPT5 MIR185 DGCR6L RTN4R RANBP1 MIR6816 COMT |
nsv515664 | Chr.22:19873352 - 19892114 on Build GRCh38 | Loss | TXNRD2 |
nsv834130 | Chr.22:19837923 - 19940567 on Build GRCh38 | Loss | C22orf29 TXNRD2 GNB1L |
esv3568260 | Chr.22:19777375 - 19881883 on Build GRCh38 | Loss | C22orf29 TBX1 TXNRD2 GNB1L |
esv3647279 | Chr.22:19719848 - 19878980 on Build GRCh38 | Gain | GP1BB C22orf29 TBX1 TXNRD2 GNB1L SEPT5-GP1BB SEPT5 |
nsv953025 | Chr.22:19869078 - 19937977 on Build GRCh38 | Deletion | TXNRD2 |
dgv7983n54 | Chr.22:19762002 - 20321870 on Build GRCh38 | Gain | LOC284865 C22orf29 TRMT2A MIR3618 TANGO2 TBX1 ZDHHC8 ARVCF MIR1306 MIR1286 GNB1L MIR4761 MIR185 LINC00896 CCDC188 DGCR6L RTN4R DGCR8 RANBP1 MIR6816 COMT TXNRD2 LOC440792 |
esv3575418 | Chr.22:18161776 - 19963846 on Build GRCh38 | Gain | UFD1L DGCR2 TBX1 PI4KAP1 DGCR5 LOC100996432 FAM230A DGCR9 TMEM191B LOC642643 C22orf39 MIR4761 LOC102725072 LOC100996415 LOC729461 MRPL40 USP18 CLTCL1 GSC2 PRODH TXNRD2 GGT3P DGCR11 SLC25A1 SEPT5-GP1BB LINC00895 DGCR10 CLDN5 GP1BB C22orf29 CDC45 DGCR14 LOC100996401 TSSK2 GNB1L SEPT5 HIRA COMT LINC01311 RIMBP3 DGCR6 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |