Assay Details
Target Gene Details
Entrez Gene ID: | 80020 |
Gene Name: | FAD dependent oxidoreductase domain containing 2 |
Gene Aliases: |
ERFAD |
Location: |
Chr.22:36487186-36507101 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FOXRED2 | NM_001102371.1 | NP_001095841.1 | ||
NM_024955.5 | NP_079231.4 | |||
XM_017028941.1 | XP_016884430.1 | |||
XM_017028942.1 | XP_016884431.1 | |||
AK025714.1 | BAB15227.1 | |||
AK026975.1 | BAB15610.1 | |||
AK091053.1 | ||||
AK225347.1 | ||||
AK315555.1 | ||||
BC015726.1 | AAH15726.1 | |||
BC027716.2 | AAH27716.1 | |||
BC040351.1 | AAH40351.1 | |||
CR456444.1 | CAG30330.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2760722 | Chr.22:36390921 - 36617980 on Build GRCh38 | Gain | CACNG2 TXN2 EIF3D FOXRED2 |
nsv520497 | Chr.22:36299896 - 36570195 on Build GRCh38 | Gain | CACNG2 TXN2 MYH9 EIF3D FOXRED2 |
nsv1060004 | Chr.22:36448479 - 36615303 on Build GRCh38 | Gain | CACNG2 TXN2 EIF3D FOXRED2 |
nsv834186 | Chr.22:36335878 - 36510765 on Build GRCh38 | Loss | TXN2 MYH9 FOXRED2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |