Assay Details
Target Gene Details
Entrez Gene ID: | 3055 |
Gene Name: | HCK proto-oncogene, Src family tyrosine kinase |
Gene Aliases: |
JTK9, p59Hck, p61Hck |
Location: |
Chr.20:32052188-32101856 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HCK | NM_001172129.1 | NP_001165600.1 | ||
NM_001172130.1 | NP_001165601.1 | |||
NM_001172131.1 | NP_001165602.1 | |||
NM_001172132.1 | NP_001165603.1 | |||
NM_001172133.1 | NP_001165604.1 | |||
NM_002110.3 | NP_002101.2 | |||
AK026432.1 | BAB15482.1 | |||
AK225819.1 | ||||
AK289896.1 | ||||
AK290928.1 | ||||
AK298726.1 | ||||
AK314335.1 | ||||
BC014435.1 | ||||
BC073748.1 | AAH73748.1 | |||
BC094847.1 | ||||
BC108930.1 | ||||
BC108931.1 | ||||
BC113854.1 | ||||
BC114463.1 | ||||
DA465978.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv817891 | Chr.20:32051627 - 32064574 on Build GRCh38 | Gain | HCK |
nsv833953 | Chr.20:32022592 - 32204778 on Build GRCh38 | Loss | TSPY26P PLAGL2 HCK CCM2L TM9SF4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |