Assay Details
Target Gene Details
Entrez Gene ID: | 160 |
Gene Name: | adaptor related protein complex 2 alpha 1 subunit |
Gene Aliases: |
ADTAA, AP2-ALPHA, CLAPA1 |
Location: |
Chr.19:49766923-49807114 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AP2A1 | NM_014203.2 | NP_055018.2 | ||
NM_130787.2 | NP_570603.2 | |||
XM_011526556.2 | XP_011524858.1 | |||
XM_011526557.2 | XP_011524859.1 | |||
AK124343.1 | ||||
AK302005.1 | ||||
AL136925.1 | CAB66859.1 | |||
BC014214.2 | AAH14214.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss | RCN3 RASIP1 ZNF114 TBC1D17 LOC101059948 EMP3 LOC101928295 MIR5088 TEAD2 FTL CGB8 CABP5 SNORD32A RPL18 CCDC155 FCGRT MIR4324 MIR4751 CGB7 DBP PTOV1-AS1 PLA2G4C-AS1 SNAR-G1 FUZ PTOV1 BCAT2 SNAR-G2 SNAR-D SNAR-A5 SNAR-A2 NTN5 SNRNP70 ATF5 ELSPBP1 IRF3 SPHK2 CGB2 FAM83E FUT1 SNAR-A11 SNAR-C5 PIH1D1 GRIN2D MIR6800 CGB5 SNORD35B CGB3 NUP62 NTF4 IL4I1 PPP1R15A SNAR-B1 CA11 ALDH16A1 TMEM143 LIN7B MIR150 SIGLEC16 SNAR-C1 GFY LIG1 PRMT1 MIR6799 CYTH2 RPS11 PNKP AP2A1 PTH2 TULP2 LHB HRC SNAR-A14 PRRG2 SNAR-A7 CCDC114 SNORD35A RUVBL2 NUCB1 CPT1C PLEKHA4 SLC17A7 GYS1 SPACA4 SNAR-C2 LMTK3 RPL13A ADM5 FLJ26850 CARD8 KCNJ14 PRR12 ZNF473 MAMSTR IZUMO1 SNAR-C3 SNAR-A1 FGF21 SNAR-C4 SNAR-A13 KCNA7 PTOV1-AS2 C19orf68 DHDH SNAR-A10 HSD17B14 GRWD1 TSKS SNAR-A6 AKT1S1 BAX LOC105447645 SCAF1 PLA2G4C SNAR-A12 MIR4750 LOC105372430 SNAR-A8 RRAS SNAR-A9 PPFIA3 BSPH1 CGB1 SIGLEC11 CARD8-AS1 NOSIP SEC1P BCL2L12 C19orf73 SNORD34 LOC100287477 SYNGR4 SNAR-A4 TRPM4 VRK3 SNAR-B2 DKKL1 FUT2 SLC6A16 SNAR-A3 SULT2B1 FLT3LG NUCB1-AS1 SNORD33 CD37 MIR4749 MED25 MIR6798 KDELR1 |
nsv470150 | Chr.19:49618821 - 49856789 on Build GRCh38 | Gain | PTOV1-AS1 ADM5 PRR12 SCAF1 AP2A1 FUZ PTOV1 MIR5088 RRAS MIR6800 PTOV1-AS2 MIR4749 PRMT1 CPT1C MED25 MIR6799 BCL2L12 TSKS IRF3 |
dgv53n68 | Chr.19:49724393 - 49943224 on Build GRCh38 | Loss | PTOV1-AS1 PNKP TBC1D17 AP2A1 MIR4750 FUZ NUP62 PTOV1 IL4I1 MIR6800 PTOV1-AS2 MIR4751 MIR4749 MED25 ATF5 MIR6799 TSKS AKT1S1 |
nsv953600 | Chr.19:49555044 - 49938043 on Build GRCh38 | Deletion | PNKP TBC1D17 SCAF1 AP2A1 MIR4750 MIR5088 RRAS MIR6800 PRRG2 MIR4751 NOSIP CPT1C BCL2L12 PTOV1-AS1 ADM5 PRR12 FUZ NUP62 PTOV1 IL4I1 PTOV1-AS2 MIR4749 PRMT1 MED25 ATF5 MIR6799 TSKS AKT1S1 IRF3 |
nsv1160655 | Chr.19:49489110 - 49841830 on Build GRCh38 | Deletion | RCN3 SCAF1 AP2A1 MIR5088 RRAS MIR6800 PRRG2 SNORD32A SNORD35B FCGRT SNORD35A NOSIP CPT1C BCL2L12 SNORD34 RPL13A PTOV1-AS1 ADM5 PRR12 FUZ MIR150 SNORD33 PRMT1 MED25 MIR6799 TSKS RPS11 IRF3 |
nsv524546 | Chr.19:49786602 - 49792888 on Build GRCh38 | Loss | AP2A1 MIR6799 |
nsv518114 | Chr.19:49789853 - 49792888 on Build GRCh38 | Loss | AP2A1 MIR6799 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |