Assay Details
Target Gene Details
Entrez Gene ID: | 374291 |
Gene Name: | NADH:ubiquinone oxidoreductase core subunit S7 |
Gene Aliases: |
CI-20, CI-20KD, MY017, PSST |
Location: |
Chr.19:1383571-1395589 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NDUFS7 | NM_024407.4 | NP_077718.3 | ||
XM_005259556.4 | XP_005259613.2 | |||
AF060512.1 | AAG43130.1 | |||
AF115969.1 | AAP97240.1 | |||
AK056061.1 | 1 | 216 | ||
AK091024.1 | 6 | 1080 | ||
AK091046.1 | 1 | 1773 | ||
AK091623.1 | 7 | 851 | ||
AK092169.1 | 6 | 1119 | BAC03820.1 | |
AK127759.1 | 6 | 2781 | BAC87117.1 | |
AK128713.1 | 5 | 1386 | BAC87587.1 | |
AK222738.1 | BAD96458.1 | |||
AK293917.1 | ||||
AK297611.1 | 6 | 923 | ||
BC001715.3 | AAH01715.2 | |||
BC005954.1 | AAH05954.1 | |||
BC111517.1 | AAI11518.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv32942 | Chr.19:1320268 - 1999205 on Build GRCh38 | Gain+Loss | MUM1 ABHD17A NDUFS7 TCF3 DAZAP1 LOC100288123 CSNK1G2 REEP6 KLF16 ADAMTSL5 SCAMP4 MBD3 GAMT ADAT3 PCSK4 C19orf25 PLK5 CSNK1G2-AS1 APC2 REXO1 MIR1909 UQCR11 RPS15 MEX3D BTBD2 ONECUT3 ATP8B3 |
nsv833705 | Chr.19:1380125 - 1544934 on Build GRCh38 | Loss | NDUFS7 PCSK4 C19orf25 PLK5 DAZAP1 APC2 REEP6 GAMT ADAMTSL5 RPS15 |
nsv482207 | Chr.19:1383884 - 1395589 on Build GRCh38 | Gain | NDUFS7 |
esv3643389 | Chr.19:1264454 - 1397379 on Build GRCh38 | Gain | CIRBP-AS1 MUM1 NDUFS7 EFNA2 C19orf24 GAMT CIRBP |
esv2758743 | Chr.19:951641 - 1544934 on Build GRCh38 | Gain+Loss | MUM1 NDUFS7 LOC102725180 DAZAP1 EFNA2 REEP6 GPX4 ADAMTSL5 CIRBP CIRBP-AS1 ARHGAP45 STK11 CBARP ARID3A GRIN3B GAMT TMEM259 ABCA7 PCSK4 LOC105372235 C19orf25 PLK5 WDR18 APC2 SBNO2 C19orf24 ATP5D RPS15 CNN2 RNU6-2 POLR2E MIDN |
nsv953942 | Chr.19:1371602 - 2057401 on Build GRCh38 | Deletion | MUM1 ABHD17A NDUFS7 TCF3 DAZAP1 LOC100288123 CSNK1G2 REEP6 KLF16 ADAMTSL5 SCAMP4 MBD3 GAMT ADAT3 PCSK4 C19orf25 PLK5 CSNK1G2-AS1 APC2 REXO1 MIR1909 UQCR11 RPS15 MEX3D BTBD2 MKNK2 ONECUT3 ATP8B3 |
esv3643396 | Chr.19:1356856 - 1396468 on Build GRCh38 | Gain | MUM1 NDUFS7 |
nsv578278 | Chr.19:1371355 - 1448181 on Build GRCh38 | Loss | MUM1 NDUFS7 DAZAP1 APC2 GAMT RPS15 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |