Assay Details
Target Gene Details
Entrez Gene ID: | 160 |
Gene Name: | adaptor related protein complex 2 alpha 1 subunit |
Gene Aliases: |
ADTAA, AP2-ALPHA, CLAPA1 |
Location: |
Chr.19:49766923-49807114 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AP2A1 | NM_014203.2 | NP_055018.2 | ||
NM_130787.2 | NP_570603.2 | |||
XM_011526556.2 | XP_011524858.1 | |||
XM_011526557.2 | XP_011524859.1 | |||
AK124343.1 | ||||
AK302005.1 | ||||
AL136925.1 | CAB66859.1 | |||
BC014214.2 | AAH14214.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss | TEAD2 SNAR-A14 SNAR-A13 BCL2L12 EMP3 MIR4751 CGB7 MED25 ELSPBP1 PLEKHA4 FUT2 FUT1 DBP PTOV1 SNAR-C2 NTN5 CA11 HRC PLA2G4C SNAR-D TULP2 SPACA4 CGB2 KCNA7 MAMSTR MIR4324 SNAR-A1 SNORD32A SNAR-A4 DKKL1 CPT1C DHDH LOC105372430 MIR6798 NUCB1 BSPH1 MIR6800 C19orf68 PRR12 C19orf73 LHB SNAR-A12 MIR5088 SNAR-B1 SNAR-A3 HSD17B14 IL4I1 FCGRT CGB8 SNORD35B LOC101059948 SNAR-B2 SIGLEC11 PPP1R15A IRF3 LOC101928295 PTOV1-AS1 TSKS ATF5 RPL18 FGF21 CABP5 PRMT1 SNAR-A5 ZNF114 ALDH16A1 SNAR-A6 SNAR-A10 CCDC114 ZNF473 SNAR-C3 CCDC155 KCNJ14 RCN3 VRK3 SNRNP70 RRAS FUZ TMEM143 SNORD35A NUCB1-AS1 BAX PPFIA3 FAM83E NOSIP LIG1 SCAF1 GRIN2D CARD8-AS1 MIR150 IZUMO1 RUVBL2 SLC17A7 SULT2B1 LOC105447645 SLC6A16 LIN7B GYS1 PLA2G4C-AS1 SNAR-C4 PTH2 AKT1S1 PRRG2 SNAR-C1 GFY MIR4750 NTF4 SNAR-C5 CGB5 SNAR-G1 MIR6799 AP2A1 CD37 SPHK2 CARD8 FTL SNAR-A11 RPL13A MIR4749 NUP62 PIH1D1 KDELR1 SNAR-A7 LMTK3 PNKP PTOV1-AS2 CYTH2 TRPM4 FLT3LG SNAR-A8 RASIP1 SIGLEC16 FLJ26850 SEC1P SNORD34 SNORD33 BCAT2 SNAR-G2 SNAR-A9 GRWD1 SNAR-A2 TBC1D17 CGB3 CGB1 RPS11 LOC100287477 ADM5 SYNGR4 |
nsv470150 | Chr.19:49618821 - 49856789 on Build GRCh38 | Gain | PRMT1 MIR6800 MIR4749 BCL2L12 PRR12 MIR5088 SCAF1 RRAS FUZ MED25 MIR6799 PTOV1-AS2 AP2A1 CPT1C IRF3 ADM5 PTOV1-AS1 TSKS PTOV1 |
dgv53n68 | Chr.19:49724393 - 49943224 on Build GRCh38 | Loss | ATF5 MIR6800 AKT1S1 MIR4749 NUP62 MIR4751 MIR4750 IL4I1 FUZ MED25 PNKP MIR6799 PTOV1-AS2 AP2A1 TBC1D17 PTOV1-AS1 TSKS PTOV1 |
nsv953600 | Chr.19:49555044 - 49938043 on Build GRCh38 | Deletion | NOSIP MIR6800 MIR4749 BCL2L12 PRR12 NUP62 MIR4751 MIR5088 SCAF1 IL4I1 MED25 PNKP PTOV1-AS2 IRF3 PTOV1-AS1 TSKS PTOV1 ATF5 PRMT1 AKT1S1 PRRG2 MIR4750 RRAS FUZ MIR6799 AP2A1 TBC1D17 CPT1C ADM5 |
nsv1160655 | Chr.19:49489110 - 49841830 on Build GRCh38 | Deletion | NOSIP RPL13A MIR6800 BCL2L12 PRR12 MIR5088 SCAF1 FCGRT MIR150 MED25 SNORD35B IRF3 PTOV1-AS1 TSKS PRMT1 PRRG2 SNORD34 SNORD33 RCN3 RRAS FUZ MIR6799 AP2A1 SNORD32A SNORD35A RPS11 CPT1C ADM5 |
nsv524546 | Chr.19:49786602 - 49792888 on Build GRCh38 | Loss | MIR6799 AP2A1 |
nsv518114 | Chr.19:49789853 - 49792888 on Build GRCh38 | Loss | MIR6799 AP2A1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |