Assay Details
Target Gene Details
Entrez Gene ID: | 126133 |
Gene Name: | aldehyde dehydrogenase 16 family member A1 |
Gene Aliases: |
- |
Location: |
Chr.19:49453216-49471050 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ALDH16A1 | NM_001145396.1 | NP_001138868.1 | ||
NM_153329.3 | NP_699160.2 | |||
AK024182.1 | 1 | 1153 | ||
AK074136.1 | BAB84962.1 | |||
AK297101.1 | ||||
AK298587.1 | ||||
AK301246.1 | ||||
BC014895.2 | AAH14895.2 | |||
DA629821.1 | ||||
DC364604.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv2518 | Chr.19:49452273 - 49465137 on Build GRCh38 | Insertion | ALDH16A1 PIH1D1 |
nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss | KCNJ14 TULP2 SNAR-A8 TSKS SNAR-A6 SNAR-A7 FAM83E C19orf68 BCAT2 TBC1D17 MIR6799 CGB8 SNAR-A11 TMEM143 FUT2 SNAR-G1 FTL SNAR-A3 SEC1P CCDC155 LIG1 LOC101928295 BCL2L12 RASIP1 PLA2G4C PTH2 PLEKHA4 SLC17A7 ADM5 MED25 NOSIP MIR6798 MIR4751 SNAR-B2 CARD8 TRPM4 SNAR-B1 CGB3 PRR12 SNORD32A RUVBL2 SPACA4 LOC101059948 SPHK2 MIR6800 SNAR-C1 RCN3 LMTK3 BAX PTOV1-AS2 SIGLEC11 SNRNP70 FLT3LG EMP3 ALDH16A1 SCAF1 RPL18 SNAR-A4 FGF21 SNAR-A2 SNAR-G2 SNAR-D KDELR1 SNAR-A12 CPT1C NUCB1-AS1 HSD17B14 PPFIA3 HRC PRMT1 CARD8-AS1 SNAR-C2 PRRG2 LOC100287477 SNAR-C3 CABP5 SNORD33 SNORD35A MIR4324 IL4I1 AP2A1 SNAR-A5 ZNF473 SIGLEC16 LOC105447645 GRWD1 DBP SNAR-C4 GRIN2D BSPH1 RPS11 CGB2 CGB1 SNAR-A10 KCNA7 CA11 FLJ26850 RRAS NTN5 PTOV1 DHDH SNAR-C5 SNAR-A13 MAMSTR SNORD35B FUT1 SLC6A16 PNKP PIH1D1 MIR4750 SNAR-A9 NUCB1 VRK3 LOC105372430 FCGRT FUZ ELSPBP1 SYNGR4 PPP1R15A LHB GFY RPL13A C19orf73 PTOV1-AS1 SNAR-A14 LIN7B CGB7 ZNF114 MIR4749 MIR150 SULT2B1 SNAR-A1 IRF3 CCDC114 CYTH2 AKT1S1 NUP62 CGB5 IZUMO1 TEAD2 GYS1 MIR5088 SNORD34 CD37 ATF5 DKKL1 PLA2G4C-AS1 NTF4 |
nsv833862 | Chr.19:49448789 - 49570676 on Build GRCh38 | Loss | SNORD32A FCGRT MIR150 SNORD33 SNORD35A RCN3 SNORD35B NOSIP RPL13A FLT3LG SNORD34 RPS11 ALDH16A1 PIH1D1 |
dgv52n68 | Chr.19:49286685 - 49456131 on Build GRCh38 | Loss | PTH2 GFY SLC6A16 TEAD2 CCDC155 SLC17A7 CD37 MIR4324 ALDH16A1 DKKL1 LOC101928295 PIH1D1 |
nsv953599 | Chr.19:49386444 - 49506743 on Build GRCh38 | Deletion | PTH2 SNORD32A MIR150 SNORD33 SNORD35A SLC17A7 SNORD35B GFY RPL13A FLT3LG SNORD34 CCDC155 RPS11 ALDH16A1 LOC101928295 PIH1D1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |