Assay Details
Target Gene Details
Entrez Gene ID: | 2217 |
Gene Name: | Fc fragment of IgG receptor and transporter |
Gene Aliases: |
FCRN, alpha-chain |
Location: |
Chr.19:49512279-49526428 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FCGRT | NM_001136019.2 | NP_001129491.1 | ||
NM_004107.4 | NP_004098.1 | |||
XM_005258657.1 | XP_005258714.1 | |||
XM_011526627.2 | XP_011524929.1 | |||
AK074734.1 | ||||
AK075510.1 | ||||
AK075532.1 | ||||
AK299124.1 | ||||
BC008734.2 | AAH08734.1 | |||
BT006828.1 | AAP35474.1 | |||
BX647163.1 | CAI46032.1 | |||
U12255.1 | AAA58958.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss | TSKS SNAR-C1 FUT1 RPL13A PLA2G4C-AS1 TRPM4 SULT2B1 SNAR-A4 PRRG2 EMP3 AKT1S1 NTF4 ELSPBP1 CCDC155 SLC6A16 LIG1 VRK3 CGB8 DBP PPP1R15A SNAR-A11 PTOV1 C19orf68 ADM5 SNORD33 SCAF1 FCGRT RRAS LOC101928295 SNORD32A CARD8-AS1 SNAR-A3 GYS1 SNAR-C2 BCL2L12 FGF21 LOC100287477 ZNF473 KCNJ14 KCNA7 ZNF114 NTN5 CGB5 PIH1D1 DHDH CGB1 LMTK3 MIR6799 GFY SNAR-A12 SNAR-G2 SNAR-C4 SNORD34 SNAR-A13 FUT2 SNRNP70 CYTH2 CGB3 IRF3 LOC101059948 LOC105447645 CGB2 NUCB1-AS1 SNAR-A8 MIR6800 ALDH16A1 SNAR-A1 LIN7B SLC17A7 PTOV1-AS1 TMEM143 MIR4324 HSD17B14 RPS11 CD37 RASIP1 LOC105372430 LHB BAX SNORD35B NOSIP NUCB1 MAMSTR PRR12 PPFIA3 MIR4750 SNAR-B1 HRC PTOV1-AS2 SNAR-A14 SNAR-A9 PLEKHA4 SNAR-C5 SNAR-G1 RPL18 IL4I1 RUVBL2 SNORD35A CARD8 FTL SPHK2 SNAR-A7 ATF5 SNAR-A6 DKKL1 GRWD1 IZUMO1 BSPH1 GRIN2D SNAR-B2 PRMT1 PTH2 TULP2 SPACA4 CCDC114 FLJ26850 FLT3LG PLA2G4C AP2A1 MED25 MIR4749 MIR6798 KDELR1 BCAT2 MIR5088 MIR150 FAM83E CGB7 CABP5 SNAR-A10 TEAD2 MIR4751 RCN3 SEC1P SNAR-C3 C19orf73 SIGLEC16 PNKP SIGLEC11 CPT1C TBC1D17 NUP62 CA11 SNAR-A2 SNAR-D FUZ SNAR-A5 SYNGR4 |
nsv833863 | Chr.19:49479177 - 49629716 on Build GRCh38 | Loss | RCN3 RPS11 SNORD35A RPL13A FLT3LG SNORD35B SNORD33 NOSIP FCGRT PRR12 PRRG2 SNORD32A SNORD34 MIR150 |
nsv833862 | Chr.19:49448789 - 49570676 on Build GRCh38 | Loss | RCN3 RPS11 SNORD35A RPL13A FLT3LG SNORD35B SNORD33 NOSIP PIH1D1 FCGRT SNORD32A ALDH16A1 SNORD34 MIR150 |
nsv1160655 | Chr.19:49489110 - 49841830 on Build GRCh38 | Deletion | TSKS RPS11 RPL13A SNORD35B NOSIP PRR12 AP2A1 MED25 PRRG2 MIR6799 SNORD34 MIR5088 MIR150 RCN3 SNORD35A ADM5 IRF3 SNORD33 CPT1C SCAF1 FCGRT RRAS SNORD32A MIR6800 PTOV1-AS1 BCL2L12 FUZ PRMT1 |
esv3644626 | Chr.19:49518636 - 49522934 on Build GRCh38 | Loss | FCGRT |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |