Assay Details
Target Gene Details
Entrez Gene ID: | 79701 |
Gene Name: | 2-oxoglutarate and iron dependent oxygenase domain containing 3 |
Gene Aliases: |
C17orf101 |
Location: |
Chr.17:82389210-82418637 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
OGFOD3 | NM_024648.2 | NP_078924.1 | ||
NM_175902.4 | NP_787098.3 | |||
NR_033265.1 | ||||
AK025875.1 | BAB15267.1 | |||
AK311057.1 | ||||
BC008897.1 | AAH08897.1 | |||
BC023602.2 | AAH23602.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1059334 | Chr.17:82277007 - 82411683 on Build GRCh38 | Gain | LOC105376790 SECTM1 OGFOD3 UTS2R LOC101929511 TEX19 CD7 |
nsv833573 | Chr.17:82275053 - 82406183 on Build GRCh38 | Loss | LOC105376790 SECTM1 OGFOD3 UTS2R LOC101929511 TEX19 CD7 |
esv2760445 | Chr.17:82267669 - 82421456 on Build GRCh38 | Gain | HEXDC LOC105376790 SECTM1 OGFOD3 UTS2R LOC101929511 TEX19 CSNK1D CD7 |
nsv428351 | Chr.17:81152631 - 83064073 on Build GRCh38 | Gain | LOC105376791 HGS ARHGDIA FN3KRP FOXK2 NPLOC4 PYCR1 TEX19 NARF TMEM105 LOC100130370 DUS1L FAAP100 LOC105371925 CSNK1D ACTG1 C17orf89 SLC16A3 MAFG LOC101929552 MCRIP1 SLC25A10 SIRT7 MIR6786 CCDC137 MAFG-AS1 TSPAN10 FSCN2 MIR6787 MIR3186 LRRC45 ARL16 ASPSCR1 LOC101929511 AATK-AS1 FASN BAHCC1 NOTUM TBCD SLC38A10 STRA13 NPB HEXDC CEP131 B3GNTL1 FN3K C17orf62 GCGR LOC105371944 PCYT2 MYADML2 PPP1R27 RAC3 ZNF750 MRPL12 PDE6G OGFOD3 ANAPC11 RAB40B CCDC57 UTS2R OXLD1 MIR4740 CD7 RFNG AATK TEPSIN SECTM1 ALYREF WDR45B GPS1 DCXR MIR4525 LINC00482 LOC105376790 P4HB SNORD134 |
nsv1067160 | Chr.17:82381853 - 82446450 on Build GRCh38 | Loss | HEXDC LOC105376790 OGFOD3 C17orf62 |
nsv828143 | Chr.17:82400094 - 82429642 on Build GRCh38 | Loss | HEXDC OGFOD3 |
nsv469561 | Chr.17:82251148 - 82472274 on Build GRCh38 | Loss | HEXDC LOC105376790 SECTM1 OGFOD3 UTS2R LOC101929511 TEX19 C17orf62 CSNK1D NARF CD7 |
nsv952394 | Chr.17:82297625 - 82408424 on Build GRCh38 | Deletion | LOC105376790 SECTM1 OGFOD3 UTS2R TEX19 CD7 |
nsv1067194 | Chr.17:82294731 - 82653129 on Build GRCh38 | Gain | HEXDC LOC105376790 FOXK2 SECTM1 OGFOD3 UTS2R TEX19 C17orf62 NARF CD7 WDR45B |
esv2762577 | Chr.17:82244836 - 82838431 on Build GRCh38 | Gain+Loss | LOC105376791 TBCD LOC101929552 HEXDC FN3KRP FOXK2 FN3K SECTM1 TEX19 C17orf62 NARF WDR45B MIR4525 ZNF750 LOC105376790 OGFOD3 RAB40B UTS2R LOC101929511 CSNK1D CD7 |
esv21913 | Chr.17:81828123 - 83103548 on Build GRCh38 | Gain+Loss | LOC105376791 ARHGDIA FN3KRP FOXK2 PYCR1 TEX19 NARF DUS1L CSNK1D SLC16A3 MAFG LOC101929552 MCRIP1 SIRT7 MAFG-AS1 MIR6787 LRRC45 ASPSCR1 LOC101929511 FASN NOTUM TBCD STRA13 NPB METRNL HEXDC B3GNTL1 FN3K C17orf62 LOC105371944 PCYT2 MYADML2 PPP1R27 RAC3 ZNF750 OGFOD3 ANAPC11 RAB40B CCDC57 UTS2R CD7 RFNG SECTM1 ALYREF WDR45B GPS1 DCXR MIR4525 LOC105376790 P4HB SNORD134 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |