Assay Details
Target Gene Details
Entrez Gene ID: | 339230 |
Gene Name: | coiled-coil domain containing 137 |
Gene Aliases: |
RaRF |
Location: |
Chr.17:81666697-81673907 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CCDC137 | NM_199287.2 | NP_954981.1 | ||
XM_011524738.2 | XP_011523040.1 | |||
XM_017024573.1 | 4 | 702 | XP_016880062.1 | |
BC009369.2 | AAH09369.1 | |||
BM548065.1 | ||||
FJ649645.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv26223 | Chr.17:80659041 - 81736487 on Build GRCh38 | Gain+Loss | LINC00482 MIR657 RPTOR C17orf89 SLC25A10 MIR3186 LOC107985021 LOC100130370 CCDC137 ACTG1 LOC100129503 BAIAP2 HGS CEP131 LOC101928855 LOC105371925 BAIAP2-AS1 TSPAN10 AATK LOC105371921 MIR1250 MIR3065 CHMP6 FSCN2 MRPL12 MIR338 LOC400627 AATK-AS1 FAAP100 ARL16 OXLD1 NPLOC4 BAHCC1 PDE6G TEPSIN TMEM105 LOC107985022 MIR4740 SLC38A10 MIR6786 |
nsv428351 | Chr.17:81152631 - 83064073 on Build GRCh38 | Gain | MAFG-AS1 OGFOD3 MAFG PPP1R27 C17orf89 SLC25A10 LOC105371944 MIR3186 FOXK2 ACTG1 LOC105376791 ZNF750 MYADML2 HGS CEP131 MIR6787 LOC105376790 UTS2R FSCN2 MRPL12 WDR45B DCXR FN3KRP DUS1L OXLD1 PYCR1 GCGR ASPSCR1 TBCD BAHCC1 PDE6G TEPSIN LOC101929552 MIR4740 SLC38A10 SLC16A3 MIR6786 LOC101929511 C17orf62 SIRT7 ANAPC11 STRA13 LINC00482 RFNG NOTUM ALYREF FN3K P4HB LRRC45 CSNK1D LOC100130370 CCDC137 CD7 RAB40B NPB LOC105371925 TSPAN10 AATK HEXDC PCYT2 TEX19 GPS1 SECTM1 AATK-AS1 FAAP100 ARL16 ARHGDIA SNORD134 MCRIP1 RAC3 MIR4525 B3GNTL1 NPLOC4 FASN CCDC57 NARF TMEM105 |
nsv470620 | Chr.17:81516973 - 82064485 on Build GRCh38 | Loss | MAFG-AS1 SIRT7 ANAPC11 MAFG STRA13 PPP1R27 RFNG NOTUM ALYREF SLC25A10 P4HB LRRC45 CCDC137 MYADML2 HGS NPB TSPAN10 PCYT2 GPS1 FSCN2 MRPL12 FAAP100 DCXR ARL16 ARHGDIA DUS1L MCRIP1 OXLD1 RAC3 PYCR1 GCGR ASPSCR1 NPLOC4 PDE6G MIR6786 |
nsv952390 | Chr.17:81637175 - 81860124 on Build GRCh38 | Deletion | TSPAN10 PPP1R27 MRPL12 SLC25A10 P4HB ARL16 MCRIP1 OXLD1 GCGR CCDC137 PDE6G HGS MIR6786 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |