Assay Details
Target Gene Details
Entrez Gene ID: | 105371932 |
Gene Name: | uncharacterized LOC105371932 |
Gene Aliases: |
- |
Location: |
Chr.17:35240043-35247472 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC105371932 | XM_017025492.1 | XP_016880981.1 |
Target Gene Details
Entrez Gene ID: | 162394 |
Gene Name: | schlafen family member 5 |
Gene Aliases: |
- |
Location: |
Chr.17:35243026-35273650 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLFN5 | NM_144975.3 | NP_659412.3 | ||
XM_005257934.3 | XP_005257991.1 | |||
AK054668.1 | BAB70788.1 | |||
AK303299.1 | ||||
AK303632.1 | ||||
BC021238.1 | AAH21238.1 | |||
BP281007.1 | ||||
BX647942.1 | ||||
CD704489.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422399 | Chr.17:35212221 - 35291582 on Build GRCh38 | Deletion | LOC105371932 SLFN5 |
esv3892998 | Chr.17:35106468 - 35295755 on Build GRCh38 | Loss | UNC45B LOC105371932 NLE1 RAD51L3-RFFL FNDC8 SLC35G3 RAD51D SLFN5 |
nsv574741 | Chr.17:35225256 - 35500115 on Build GRCh38 | Loss | SLFN13 SLFN12L LOC105371933 LOC105371932 SLFN11 SLFN12 SLFN5 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |