Assay Details
Target Gene Details
Entrez Gene ID: | 79701 |
Gene Name: | 2-oxoglutarate and iron dependent oxygenase domain containing 3 |
Gene Aliases: |
C17orf101 |
Location: |
Chr.17:82389210-82418637 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
OGFOD3 | NM_024648.2 | NP_078924.1 | ||
NM_175902.4 | NP_787098.3 | |||
NR_033265.1 | ||||
AK025875.1 | BAB15267.1 | |||
AK311057.1 | ||||
BC008897.1 | AAH08897.1 | |||
BC023602.2 | AAH23602.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1059334 | Chr.17:82277007 - 82411683 on Build GRCh38 | Gain | CD7 LOC105376790 SECTM1 UTS2R TEX19 LOC101929511 OGFOD3 |
nsv833573 | Chr.17:82275053 - 82406183 on Build GRCh38 | Loss | CD7 LOC105376790 SECTM1 UTS2R TEX19 LOC101929511 OGFOD3 |
esv2760445 | Chr.17:82267669 - 82421456 on Build GRCh38 | Gain | CD7 LOC105376790 SECTM1 UTS2R TEX19 CSNK1D HEXDC LOC101929511 OGFOD3 |
nsv428351 | Chr.17:81152631 - 83064073 on Build GRCh38 | Gain | LOC105376790 MCRIP1 PDE6G HGS TBCD UTS2R FASN MIR6787 DUS1L PYCR1 NOTUM MAFG-AS1 SLC38A10 ARL16 MIR4525 CCDC57 P4HB LOC105371944 OXLD1 HEXDC ALYREF MIR4740 MYADML2 RAB40B STRA13 PCYT2 GPS1 CCDC137 ANAPC11 MIR3186 SNORD134 TEPSIN RFNG LOC101929511 TSPAN10 NARF SLC25A10 C17orf89 SIRT7 MRPL12 LOC100130370 ZNF750 AATK-AS1 FN3K TEX19 B3GNTL1 MIR6786 TMEM105 FN3KRP CD7 C17orf62 WDR45B SECTM1 AATK CEP131 ACTG1 CSNK1D RAC3 LINC00482 DCXR SLC16A3 MAFG PPP1R27 LOC105371925 FOXK2 LOC105376791 LRRC45 NPLOC4 ARHGDIA FAAP100 GCGR ASPSCR1 LOC101929552 NPB FSCN2 BAHCC1 OGFOD3 |
nsv1067160 | Chr.17:82381853 - 82446450 on Build GRCh38 | Loss | LOC105376790 C17orf62 HEXDC OGFOD3 |
nsv469561 | Chr.17:82251148 - 82472274 on Build GRCh38 | Loss | CD7 LOC105376790 C17orf62 SECTM1 UTS2R TEX19 CSNK1D HEXDC LOC101929511 NARF OGFOD3 |
nsv952394 | Chr.17:82297625 - 82408424 on Build GRCh38 | Deletion | CD7 LOC105376790 SECTM1 UTS2R TEX19 OGFOD3 |
nsv1067194 | Chr.17:82294731 - 82653129 on Build GRCh38 | Gain | CD7 LOC105376790 C17orf62 WDR45B FOXK2 SECTM1 UTS2R TEX19 HEXDC NARF OGFOD3 |
esv2762577 | Chr.17:82244836 - 82838431 on Build GRCh38 | Gain+Loss | LOC105376790 RAB40B ZNF750 FOXK2 LOC105376791 FN3K TBCD UTS2R TEX19 FN3KRP CD7 C17orf62 WDR45B MIR4525 SECTM1 LOC101929552 CSNK1D HEXDC LOC101929511 NARF OGFOD3 |
esv21913 | Chr.17:81828123 - 83103548 on Build GRCh38 | Gain+Loss | LOC105376790 MCRIP1 TBCD UTS2R FASN MIR6787 DUS1L PYCR1 NOTUM MAFG-AS1 MIR4525 CCDC57 P4HB LOC105371944 HEXDC ALYREF MYADML2 RAB40B STRA13 PCYT2 GPS1 ANAPC11 SNORD134 RFNG LOC101929511 NARF SIRT7 ZNF750 FN3K TEX19 B3GNTL1 FN3KRP CD7 C17orf62 WDR45B SECTM1 CSNK1D METRNL RAC3 DCXR SLC16A3 MAFG PPP1R27 FOXK2 LOC105376791 LRRC45 ARHGDIA ASPSCR1 LOC101929552 NPB OGFOD3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |