Assay Details
Target Gene Details
Entrez Gene ID: | 10458 |
Gene Name: | BAI1 associated protein 2 |
Gene Aliases: |
BAP2, FLAF3, IRSP53 |
Location: |
Chr.17:81035147-81117432 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv26223 | Chr.17:80659041 - 81736487 on Build GRCh38 | Gain+Loss | SLC25A10 FAAP100 TMEM105 LOC100129503 LOC101928855 LOC105371925 LINC00482 LOC107985021 BAIAP2-AS1 AATK MIR6786 OXLD1 NPLOC4 LOC105371921 TSPAN10 MIR4740 BAHCC1 CHMP6 TEPSIN LOC107985022 LOC100130370 ACTG1 MIR1250 CEP131 HGS AATK-AS1 C17orf89 MIR3065 LOC400627 MRPL12 BAIAP2 PDE6G RPTOR MIR657 ARL16 SLC38A10 MIR338 MIR3186 FSCN2 CCDC137 |
nsv833564 | Chr.17:81014713 - 81164054 on Build GRCh38 | Loss | BAIAP2-AS1 BAIAP2 AATK MIR1250 MIR657 MIR338 MIR3065 |
nsv470619 | Chr.17:81032233 - 81349843 on Build GRCh38 | Loss | MIR1250 TMEM105 CEP131 AATK-AS1 LOC105371925 C17orf89 MIR3065 LINC00482 BAIAP2-AS1 BAIAP2 AATK MIR657 SLC38A10 MIR338 TEPSIN |
nsv576169 | Chr.17:81030816 - 81043967 on Build GRCh38 | Loss | BAIAP2-AS1 BAIAP2 |
dgv3292n100 | Chr.17:81019582 - 81204920 on Build GRCh38 | Gain | BAIAP2-AS1 BAIAP2 AATK MIR1250 MIR657 CEP131 MIR338 AATK-AS1 MIR3065 |
nsv952388 | Chr.17:81034701 - 81137300 on Build GRCh38 | Deletion | BAIAP2-AS1 BAIAP2 AATK MIR1250 MIR657 MIR338 MIR3065 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |