Assay Details
Target Gene Details
Entrez Gene ID: | 10458 |
Gene Name: | BAI1 associated protein 2 |
Gene Aliases: |
BAP2, FLAF3, IRSP53 |
Location: |
Chr.17:81035147-81117432 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv26223 | Chr.17:80659041 - 81736487 on Build GRCh38 | Gain+Loss | HGS SLC38A10 CHMP6 MIR1250 LOC100130370 LOC400627 C17orf89 FAAP100 BAHCC1 LOC100129503 BAIAP2 OXLD1 PDE6G TMEM105 MIR338 BAIAP2-AS1 FSCN2 CEP131 AATK SLC25A10 ARL16 MIR4740 LOC107985021 LOC105371921 LINC00482 LOC107985022 MIR6786 TSPAN10 LOC101928855 AATK-AS1 LOC105371925 MIR3186 CCDC137 MRPL12 ACTG1 NPLOC4 TEPSIN MIR657 MIR3065 RPTOR |
nsv833564 | Chr.17:81014713 - 81164054 on Build GRCh38 | Loss | BAIAP2 MIR1250 MIR657 MIR338 BAIAP2-AS1 MIR3065 AATK |
nsv470619 | Chr.17:81032233 - 81349843 on Build GRCh38 | Loss | SLC38A10 LINC00482 MIR1250 C17orf89 AATK-AS1 LOC105371925 BAIAP2 TEPSIN MIR657 TMEM105 MIR338 BAIAP2-AS1 CEP131 MIR3065 AATK |
dgv3292n100 | Chr.17:81019582 - 81204920 on Build GRCh38 | Gain | BAIAP2 MIR1250 MIR657 MIR338 BAIAP2-AS1 AATK-AS1 CEP131 MIR3065 AATK |
nsv952388 | Chr.17:81034701 - 81137300 on Build GRCh38 | Deletion | BAIAP2 MIR1250 MIR657 MIR338 BAIAP2-AS1 MIR3065 AATK |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |