Assay Details
Target Gene Details
Entrez Gene ID: | 10458 |
Gene Name: | BAI1 associated protein 2 |
Gene Aliases: |
BAP2, FLAF3, IRSP53 |
Location: |
Chr.17:81035147-81117432 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv26223 | Chr.17:80659041 - 81736487 on Build GRCh38 | Gain+Loss | BAIAP2 C17orf89 PDE6G LOC107985021 TMEM105 LOC105371921 FSCN2 AATK-AS1 HGS MRPL12 MIR4740 ACTG1 LOC105371925 FAAP100 TSPAN10 SLC25A10 SLC38A10 LOC100130370 MIR338 CCDC137 MIR3186 MIR1250 LINC00482 ARL16 BAHCC1 LOC107985022 CHMP6 MIR657 BAIAP2-AS1 MIR3065 TEPSIN OXLD1 MIR6786 RPTOR CEP131 LOC101928855 LOC400627 NPLOC4 AATK LOC100129503 |
nsv833564 | Chr.17:81014713 - 81164054 on Build GRCh38 | Loss | BAIAP2 MIR657 MIR338 BAIAP2-AS1 MIR3065 MIR1250 AATK |
nsv470619 | Chr.17:81032233 - 81349843 on Build GRCh38 | Loss | BAIAP2 SLC38A10 MIR338 MIR1250 C17orf89 LINC00482 TMEM105 MIR657 AATK-AS1 BAIAP2-AS1 MIR3065 TEPSIN CEP131 LOC105371925 AATK |
nsv576169 | Chr.17:81030816 - 81043967 on Build GRCh38 | Loss | BAIAP2 BAIAP2-AS1 |
dgv3292n100 | Chr.17:81019582 - 81204920 on Build GRCh38 | Gain | BAIAP2 MIR657 AATK-AS1 MIR338 BAIAP2-AS1 MIR3065 MIR1250 CEP131 AATK |
nsv952388 | Chr.17:81034701 - 81137300 on Build GRCh38 | Deletion | BAIAP2 MIR657 MIR338 BAIAP2-AS1 MIR3065 MIR1250 AATK |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |