Assay Details
Target Gene Details
Entrez Gene ID: | 6530 |
Gene Name: | solute carrier family 6 member 2 |
Gene Aliases: |
NAT1, NET, NET1, SLC6A5 |
Location: |
Chr.16:55655604-55706192 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC6A2 | NM_001043.3 | NP_001034.1 | ||
NM_001172501.1 | NP_001165972.1 | |||
NM_001172504.1 | NP_001165975.1 | |||
XM_006721263.2 | XP_006721326.1 | |||
XM_011523295.1 | XP_011521597.1 | |||
XM_011523296.1 | XP_011521598.1 | |||
XM_011523297.2 | XP_011521599.1 | |||
XM_011523298.1 | XP_011521600.1 | |||
AK312793.1 | ||||
AU132072.1 | ||||
M65105.1 | AAA59943.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3571298 | Chr.16:55605227 - 55802076 on Build GRCh38 | Gain | SLC6A2 CES1P2 CES1P1 |
esv2758649 | Chr.16:55635484 - 55949678 on Build GRCh38 | Gain | SLC6A2 CES1P2 CES1P1 CES5A CES1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |