Assay Details
Target Gene Details
Entrez Gene ID: | 146167 |
Gene Name: | solute carrier family 38 member 8 |
Gene Aliases: |
FVH2 |
Location: |
Chr.16:84009667-84043372 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC38A8 | NM_001080442.2 | NP_001073911.1 | ||
XM_017022946.1 | XP_016878435.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv528396 | Chr.16:83937637 - 84050522 on Build GRCh38 | Loss | SLC38A8 OSGIN1 LOC105371371 NECAB2 |
nsv1065790 | Chr.16:83830329 - 84243597 on Build GRCh38 | Loss | SLC38A8 MLYCD OSGIN1 DNAAF1 KCNG4 LOC105371371 ADAD2 TAF1C LOC729887 HSDL1 NECAB2 MBTPS1 |
nsv1909 | Chr.16:84031468 - 84076703 on Build GRCh38 | Deletion | SLC38A8 MBTPS1 |
esv3892925 | Chr.16:84022293 - 84049179 on Build GRCh38 | Loss | SLC38A8 |
dgv3056n100 | Chr.16:83923502 - 84039073 on Build GRCh38 | Loss | SLC38A8 OSGIN1 LOC105371371 NECAB2 |
nsv1067050 | Chr.16:83990319 - 84039073 on Build GRCh38 | Loss | SLC38A8 NECAB2 |
nsv457595 | Chr.16:84013110 - 84091665 on Build GRCh38 | Gain | SLC38A8 MBTPS1 |
nsv1059227 | Chr.16:84029323 - 84233403 on Build GRCh38 | Loss | SLC38A8 DNAAF1 KCNG4 ADAD2 TAF1C LOC729887 HSDL1 MBTPS1 |
esv3892924 | Chr.16:84014717 - 84091665 on Build GRCh38 | Gain | SLC38A8 MBTPS1 |
esv2761930 | Chr.16:84007420 - 84312461 on Build GRCh38 | Gain | LOC105376777 SLC38A8 WFDC1 DNAAF1 KCNG4 ADAD2 TAF1C LOC729887 HSDL1 MBTPS1 |
dgv546e214 | Chr.16:84011032 - 84095132 on Build GRCh38 | Gain | SLC38A8 MBTPS1 |
dgv5261n54 | Chr.16:84004904 - 84047164 on Build GRCh38 | Loss | SLC38A8 |
nsv527489 | Chr.16:84030777 - 84050522 on Build GRCh38 | Loss | SLC38A8 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |