Assay Details
Target Gene Details
Entrez Gene ID: | 51005 |
Gene Name: | amidohydrolase domain containing 2 |
Gene Aliases: |
CGI-14 |
Location: |
Chr.16:2520351-2531414 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AMDHD2 | NM_001145815.1 | NP_001139287.1 | ||
NM_015944.3 | NP_057028.2 | |||
XM_017023262.1 | XP_016878751.1 | |||
XM_017023263.1 | XP_016878752.1 | |||
XM_017023264.1 | XP_016878753.1 | |||
XM_017023265.1 | XP_016878754.1 | |||
XM_017023266.1 | XP_016878755.1 | |||
XM_017023267.1 | XP_016878756.1 | |||
AB209351.1 | 1 | 1545 | BAD92588.1 | |
AF132948.1 | AAD27723.1 | |||
AK296877.1 | ||||
AK300747.1 | ||||
BC018734.1 | AAH18734.1 | |||
CN362870.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication | RNPS1 MIR6768 STUB1 PDIA2 LOC100287175 SLC9A3R2 KCTD5 C1QTNF8 MIR3178 GNG13 MIR5587 LOC105371045 LOC106660606 RAB11FIP3 FAM234A C16orf59 UNKL MIR3177 TPSB2 MLST8 SNHG19 NME4 GFER CHTF18 LOC101929280 MIR6511B1 HN1L METRN LMF1 CEMP1 PKD1 MIR940 NHLRC4 CRAMP1 TPSG1 POLR3K E4F1 RHBDL1 NTN3 RAB40C JMJD8 MCRIP2 SNRNP25 LOC100134368 RGS11 RHOT2 PRR25 FAM173A MIR4717 MRPL28 CAPN15 CCDC78 SNORA10 UBE2I ABCA17P MSLN HBA1 LOC107987233 METTL26 HAGHL NME3 LOC105371047 CCDC154 SNORA78 CLCN7 MIR3180-5 LOC652276 WASIR2 ARHGDIG TELO2 PIGQ EME2 RPL23AP5 LMF1-AS1 ATP6V0C SNHG9 NPRL3 PRSS29P HBA2 LINC00235 PDPK1 RNF151 RPL3L CCNF BRICD5 WDR24 WFIKKN1 HBZ LOC729652 RHBDF1 RAB26 DDX11L10 AMDHD2 RPS2 NUBP2 GNPTG AXIN1 TSC2 TRAF7 IGFALS LUC7L MIR3176 CASKIN1 LOC105371049 SSTR5-AS1 MEIOB HS3ST6 SSTR5 MIR6767 ECI1 LOC105371184 DNASE1L2 PRSS27 SNORA64 MIR1225 SNORD60 MIR662 ABCA3 TSR3 LOC105371038 LOC105371046 MIR3677 MAPK8IP3 PGP WDR90 RPUSD1 FBXL16 IFT140 NTHL1 C16orf91 LINC00254 TPSD1 TMEM8A HAGH TMEM204 FLJ42627 BAIAP3 SOX8 FAHD1 MRPS34 SYNGR3 CACNA1H MPG MSRB1 HBQ1 TBC1D24 MIR4516 ERVK13-1 TPSAB1 NARFL ZNF598 PTX4 SPSB3 MIR6859-4 NOXO1 NPW PRR35 HBM TBL3 DECR2 NDUFB10 LOC107984876 |
esv2760067 | Chr.16:2486824 - 2820535 on Build GRCh38 | Gain+Loss | PDPK1 SRRM2-AS1 TBC1D24 PRSS33 ERVK13-1 SRRM2 LOC652276 CEMP1 KCTD5 FLJ42627 PRSS41 PRSS27 AMDHD2 MIR3178 PRSS21 TCEB2 ATP6V0C |
nsv833122 | Chr.16:2501717 - 2663937 on Build GRCh38 | Loss | CEMP1 FLJ42627 PDPK1 AMDHD2 MIR3178 TBC1D24 ERVK13-1 ATP6V0C LOC652276 |
nsv1052260 | Chr.16:2074546 - 2629655 on Build GRCh38 | Gain | NTN3 PDPK1 RNPS1 CCNF MIR6768 BRICD5 MIR4717 LOC729652 RAB26 AMDHD2 MIR3178 LOC106660606 ABCA17P C16orf59 TSC2 TRAF7 MLST8 SNHG19 TBC1D24 MIR4516 CASKIN1 LOC105371049 MIR3180-5 LOC652276 MIR6511B1 CEMP1 PKD1 MIR6767 ECI1 DNASE1L2 MIR940 MIR1225 SNORD60 ABCA3 MIR3677 ATP6V0C PGP E4F1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |