Assay Details
Target Gene Details
Entrez Gene ID: | 283692 |
Gene Name: | CPEB1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.15:82647770-82692820 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CPEB1-AS1 | NR_046096.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3637058 | Chr.15:82590287 - 82726611 on Build GRCh38 | Gain | LOC338963 AP3B2 CPEB1-AS1 CPEB1 ACTG1P17 |
dgv2637n100 | Chr.15:82568764 - 82803755 on Build GRCh38 | Gain | LOC338963 FSD2 AP3B2 CPEB1-AS1 CPEB1 ACTG1P17 SNHG21 SCARNA15 |
nsv817702 | Chr.15:82630710 - 82788672 on Build GRCh38 | Loss | LOC338963 FSD2 AP3B2 CPEB1-AS1 CPEB1 ACTG1P17 SNHG21 SCARNA15 |
esv34318 | Chr.15:82614804 - 82784743 on Build GRCh38 | Loss | LOC338963 FSD2 AP3B2 CPEB1-AS1 CPEB1 ACTG1P17 SNHG21 SCARNA15 |
esv2674021 | Chr.15:82628241 - 82789040 on Build GRCh38 | Deletion | LOC338963 FSD2 AP3B2 CPEB1-AS1 CPEB1 ACTG1P17 SNHG21 SCARNA15 |