Assay Details
Target Gene Details
Entrez Gene ID: | 64506 |
Gene Name: | cytoplasmic polyadenylation element binding protein 1 |
Gene Aliases: |
CPE-BP1, CPEB, CPEB-1, h-CPEB, hCPEB-1 |
Location: |
Chr.15:82543201-82648016 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CPEB1 | NM_001079533.1 | NP_001073001.1 | ||
NM_001079534.1 | NP_001073002.1 | |||
NM_001079535.1 | NP_001073003.1 | |||
NM_001288819.1 | NP_001275748.1 | |||
NM_001288820.1 | NP_001275749.1 | |||
NM_030594.4 | NP_085097.3 | |||
AF329402.1 | AAK01239.1 | |||
AF329403.1 | AAK01240.1 | |||
AK023265.1 | BAB14496.1 | |||
AK294280.1 | ||||
AK300105.1 | ||||
AK301728.1 | ||||
BC035348.1 | AAH35348.1 | |||
BC050629.1 | AAH50629.1 | |||
BX649159.1 | ||||
DA340952.1 | ||||
DA705313.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv483125 | Chr.15:82448179 - 82617091 on Build GRCh38 | Loss | GOLGA6L17P RPS17 CPEB1 GOLGA2P10 |
nsv833071 | Chr.15:82454948 - 82641759 on Build GRCh38 | Gain | GOLGA6L17P RPS17 CPEB1 GOLGA2P10 |
nsv1035899 | Chr.15:82439721 - 82646980 on Build GRCh38 | Gain | GOLGA6L17P RPS17 LOC102724034 CPEB1 GOLGA2P10 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |