Assay Details
Target Gene Details
Entrez Gene ID: | 6315 |
Gene Name: | ATXN8 opposite strand (non-protein coding) |
Gene Aliases: |
KLHL1AS, NCRNA00003, SCA8 |
Location: |
Chr.13:70107213-70139753 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ATXN8OS | NR_002717.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1051815 | Chr.13:69996822 - 70161932 on Build GRCh38 | Gain | KLHL1 ATXN8OS |
nsv1053979 | Chr.13:70079770 - 70208426 on Build GRCh38 | Loss | KLHL1 ATXN8OS |
nsv455967 | Chr.13:70066513 - 70131876 on Build GRCh38 | Loss | KLHL1 ATXN8OS |