Assay Details
Target Gene Details
Entrez Gene ID: | 23457 |
Gene Name: | ATP binding cassette subfamily B member 9 |
Gene Aliases: |
EST122234, TAPL |
Location: |
Chr.12:122918708-122975160 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ABCB9 | NM_001243014.1 | NP_001229943.1 | ||
AB112583.1 | BAC98410.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv22n68 | Chr.12:122911380 - 123104743 on Build GRCh38 | Loss | ARL6IP4 LOC100507091 MIR4304 ABCB9 PITPNM2 OGFOD2 |
nsv517200 | Chr.12:122827504 - 123169951 on Build GRCh38 | Loss | ARL6IP4 MPHOSPH9 LOC100507091 MIR4304 HIP1R VPS37B ABCB9 PITPNM2 OGFOD2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |