Assay Details
Target Gene Details
Entrez Gene ID: | 374378 |
Gene Name: | polypeptide N-acetylgalactosaminyltransferase 18 |
Gene Aliases: |
GALNACT18, GALNT15, GALNTL4, GalNAc-T15, GalNAc-T18 |
Location: |
Chr.11:11270874-11622045 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GALNT18 | NM_198516.2 | NP_940918.2 | ||
XM_006718224.2 | XP_006718287.1 | |||
XM_011520068.2 | XP_011518370.1 | |||
AB078147.1 | BAD93179.1 | |||
AJ626724.1 | CAF25035.1 | |||
AK055111.1 | ||||
BC037341.2 | AAH37341.3 | |||
BC060864.1 | AAH60864.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832064 | Chr.11:11128217 - 11316356 on Build GRCh38 | Gain | GALNT18 |
nsv483000 | Chr.11:10678454 - 12678453 on Build GRCh38 | Loss | GALNT18 DKK3 TEAD1 CSNK2A3 MIR6124 MICAL2 LOC105376545 ZBED5 ZBED5-AS1 CTR9 EIF4G2 LOC105376554 MIR4299 PARVA LOC101928053 MICALCL USP47 MRVI1 SNORD97 MIR8070 |
esv34098 | Chr.11:11248654 - 11450079 on Build GRCh38 | Loss | GALNT18 CSNK2A3 |
nsv832065 | Chr.11:11279220 - 11459680 on Build GRCh38 | Gain | GALNT18 CSNK2A3 |
esv32920 | Chr.11:11194328 - 11359745 on Build GRCh38 | Gain+Loss | GALNT18 CSNK2A3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |